Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGLEC5
Entrez Gene ID
8778
Gene Name
sialic acid binding Ig like lectin 5
Gene Aliases
CD170, CD33L2, OB-BP2, OBBP2, SIGLEC-5
Chromosome Location
Chr.19: 51610967 - 51630474 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003830.3 NP_003821.1 9 - 9 1653 87
XM_011527438.2 XP_011525740.1 8 - 8 1525 87
XM_017027419.1 XP_016882908.1 9 - 9 1634 87
GenBank mRNA AF170484.1 - 9 - 9 1550 87
AK225753.1 - 9 - 9 1574 87
BC029896.1 - 9 - 9 1555 87
U71383.1 - 9 - 9 1656 87

View Full Product Details