Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIP
Entrez Gene ID
2695
Gene Name
gastric inhibitory polypeptide
Gene Aliases
-
Chromosome Location
Chr.17: 48958556 - 48968593 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004123.2 NP_004114.1 2 - 2 148 75
GenBank mRNA BC069100.1 - 2 - 2 148 75
BC069663.1 - 2 - 2 148 75
BC069686.1 - 2 - 2 148 75
BC069746.1 - 2 - 2 148 75
BC096146.1 - 2 - 2 147 75
BC096147.1 - 2 - 2 147 75
BC096148.3 - 2 - 2 147 75
BC096149.3 - 2 - 2 147 75
E02006.1 - 2 - 2 150 75
M18185.1 - 2 - 2 148 75

View Full Product Details