The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004690.3 | NP_004681.1 | 8 - 8 | 4676 | 91 |
NR_073033.1 | 9 - 9 | 4582 | 91 | ||
XM_017011474.1 | XP_016866963.1 | 9 - 9 | 4617 | 91 | |
XM_017011475.1 | XP_016866964.1 | 9 - 9 | 4537 | 91 | |
XM_017011476.1 | XP_016866965.1 | 10 - 10 | 4685 | 91 | |
XM_017011478.1 | XP_016866967.1 | 9 - 9 | 4595 | 91 | |
XM_017011480.1 | XP_016866969.1 | 8 - 8 | 4314 | 91 | |
XM_017011481.1 | XP_016866970.1 | 7 - 7 | 4360 | 91 | |
GenBank mRNA | AF164041.1 | - | 7 - 7 | 4161 | 91 |