The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004933.2 | NP_004924.1 | 14 - 14 | 2809 | 84 |
GenBank mRNA | AW452260.1 | - | 1 - 1 | 84 |