The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042494.1 | NP_001035959.1 | 26 - 26 | 4176 | 78 |
NM_001042495.1 | NP_001035960.1 | 26 - 26 | 3851 | 78 | |
NM_001042496.1 | NP_001035961.1 | 26 - 26 | 4242 | 78 | |
NM_001042497.1 | NP_001035962.1 | 24 - 24 | 3945 | 78 | |
NM_005135.2 | NP_005126.1 | 25 - 25 | 3815 | 78 | |
NM_133647.1 | NP_598408.1 | 25 - 25 | 3990 | 78 | |
XM_006720793.3 | XP_006720856.1 | 24 - 24 | 4725 | 78 | |
GenBank mRNA | AF108831.1 | - | 25 - 25 | 3724 | 78 |
AK128133.1 | - | 24 - 24 | 3498 | 78 | |
AL117500.1 | - | 22 - 22 | 3418 | 78 | |
BC051744.1 | - | 26 - 26 | 4208 | 78 | |
BC070107.1 | - | 26 - 26 | 3846 | 78 | |
BX648195.1 | - | 25 - 25 | 4862 | 78 |