The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005744.3 | NP_005735.2 | 12 - 12 | 1676 | 60 |
XM_011521438.2 | XP_011519740.1 | 11 - 11 | 1652 | 60 | |
XM_011521439.2 | XP_011519741.1 | 13 - 13 | 1190 | 60 | |
GenBank mRNA | AB014774.1 | - | 3 - 3 | 236 | 60 |
AB209432.1 | - | 13 - 13 | 1305 | 60 | |
AF072832.1 | - | 12 - 12 | 1362 | 60 | |
AJ009771.1 | - | 5 - 5 | 472 | 60 | |
AJ130976.1 | - | 12 - 12 | 1080 | 60 | |
AJ243190.1 | - | 12 - 12 | 1676 | 60 | |
AK312715.1 | - | 12 - 12 | 1675 | 60 | |
BC051877.1 | - | 12 - 12 | 1611 | 60 |