The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170714.1 | NP_001164185.1 | 8 - 8 | 3156 | 114 |
NM_001170715.1 | NP_001164186.1 | 7 - 7 | 2861 | 114 | |
NM_001170716.1 | NP_001164187.1 | 7 - 7 | 2818 | 114 | |
NM_001170717.1 | NP_001164188.1 | 7 - 7 | 2934 | 114 | |
NM_001170718.1 | NP_001164189.1 | 7 - 7 | 2905 | 114 | |
NM_001170719.1 | NP_001164190.1 | 7 - 7 | 2995 | 114 | |
NM_001170720.1 | NP_001164191.1 | 8 - 8 | 2566 | 114 | |
NM_001170721.1 | NP_001164192.1 | 6 - 6 | 3093 | 114 | |
NM_014567.3 | NP_055382.2 | 7 - 7 | 2880 | 114 | |
XM_005256258.4 | XP_005256315.1 | 7 - 7 | 3080 | 114 | |
XM_005256259.4 | XP_005256316.1 | 7 - 7 | 5992 | 114 | |
XM_005256260.4 | XP_005256317.1 | 7 - 7 | 3001 | 114 | |
XM_011523475.2 | XP_011521777.1 | 7 - 7 | 3029 | 114 | |
XM_017023897.1 | XP_016879386.1 | 7 - 7 | 3587 | 114 | |
XM_017023898.1 | XP_016879387.1 | 6 - 6 | 3497 | 114 | |
XM_017023899.1 | XP_016879388.1 | 5 - 5 | 3308 | 114 | |
GenBank mRNA | AB040024.1 | - | 7 - 7 | 2875 | 114 |
AF218451.1 | - | 7 - 7 | 2858 | 114 | |
AJ242987.2 | - | 7 - 7 | 2855 | 114 | |
AK026121.1 | - | 3 - 3 | 1364 | 114 | |
AK027608.1 | - | 7 - 7 | 2905 | 114 | |
AK074958.1 | - | 6 - 6 | 2290 | 114 | |
AK124526.1 | - | 7 - 7 | 4300 | 114 | |
AK124815.1 | - | 7 - 7 | 2864 | 114 | |
AK124844.1 | - | 6 - 6 | 2671 | 114 | |
AK125719.1 | - | 3 - 3 | 2339 | 114 | |
AK293808.1 | - | 8 - 8 | 3156 | 114 | |
AK302617.1 | - | 8 - 8 | 2566 | 114 | |
BC014402.1 | - | 3 - 3 | 1522 | 114 | |
BC062556.1 | - | 6 - 6 | 2735 | 114 | |
BU730553.1 | - | 1 - 1 | 114 |