Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EAPP
Entrez Gene ID
55837
Gene Name
E2F associated phosphoprotein
Gene Aliases
BM036, C14orf11
Chromosome Location
Chr.14: 34515929 - 34539737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318916.1 NP_001305845.1 5 - 5 1053 103
NM_018453.3 NP_060923.2 6 - 6 1171 103
XM_011536958.1 XP_011535260.1 6 - 6 1449 103
GenBank mRNA AF217512.1 - 6 - 6 1161 103
AK000585.1 - 6 - 6 1135 103
AK225467.1 - 6 - 6 1125 102
AK225477.1 - 6 - 6 1135 103
AK225991.1 - 6 - 6 1135 103
AY157300.1 - 6 - 6 1157 103
AY157301.1 - 6 - 6 1157 103
AY869694.1 - 6 - 6 1094 103
BC001245.1 - 6 - 6 1144 103

View Full Product Details