The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083615.3 | NP_001077084.2 | 34 - 34 | 4840 | 79 |
NM_138995.4 | NP_620482.3 | 35 - 35 | 4921 | 79 | |
NR_045682.1 | 36 - 36 | 5043 | 79 | ||
NR_045683.1 | 36 - 36 | 4952 | 79 | ||
NR_045684.1 | 35 - 35 | 4871 | 79 | ||
XM_006712299.3 | XP_006712362.1 | 35 - 35 | 4809 | 79 | |
XM_011510654.2 | XP_011508956.1 | 36 - 36 | 4917 | 79 | |
XM_011510655.2 | XP_011508957.1 | 34 - 34 | 4728 | 79 | |
XM_011510656.2 | XP_011508958.1 | 30 - 30 | 4292 | 79 | |
XM_011510657.2 | XP_011508959.1 | 29 - 29 | 4164 | 79 | |
XM_011510658.2 | XP_011508960.1 | 30 - 30 | 4473 | 79 | |
GenBank mRNA | AF369908.1 | - | 36 - 36 | 4952 | 79 |
AF391554.1 | - | 35 - 35 | 4871 | 79 | |
AF391555.1 | - | 35 - 35 | 4921 | 79 | |
AF391556.1 | - | 34 - 34 | 4840 | 79 | |
BX647895.1 | - | 28 - 28 | 4006 | 79 |