The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144606.5 | NP_653207.1 | 8 - 8 | 1556 | 68 |
GenBank mRNA | AL831885.1 | - | 2 - 2 | 895 | 68 |
BC015687.2 | - | 8 - 8 | 1507 | 68 |