Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL23A1
Entrez Gene ID
91522
Gene Name
collagen type XXIII alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.5: 178237616 - 178590555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173465.3 NP_775736.2 29 - 29 3010 85
XM_006714933.3 XP_006714996.1 30 - 30 2818 85
XM_006714934.3 XP_006714997.1 29 - 29 2791 85
XM_006714935.3 XP_006714998.1 29 - 29 2782 85
XM_006714936.3 XP_006714999.1 29 - 29 2755 85
XM_011534688.2 XP_011532990.1 29 - 29 2791 85
XM_011534689.2 XP_011532991.1 29 - 29 2764 85
XM_011534690.2 XP_011532992.1 28 - 28 2764 85
XM_011534691.2 XP_011532993.1 26 - 26 2710 85
XM_011534692.2 XP_011532994.1 30 - 30 3843 85
GenBank mRNA AW014891.1 - 1 - 1 85
AY158895.1 - 29 - 29 3009 85
AY898961.1 - 29 - 29 3009 85

View Full Product Details