The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145008.1 | NP_001138480.1 | 5 - 5 | 1092 | 115 |
NM_001145009.1 | NP_001138481.1 | 5 - 5 | 1248 | 115 | |
NM_007048.5 | NP_008979.3 | 5 - 5 | 1248 | 115 | |
NM_194441.2 | NP_919423.1 | 5 - 5 | 1078 | 115 | |
XM_005248833.2 | XP_005248890.1 | 5 - 5 | 1216 | 115 | |
XM_005248834.3 | XP_005248891.1 | 5 - 5 | 1243 | 115 | |
XM_006714983.2 | XP_006715046.1 | 4 - 4 | 953 | 115 | |
GenBank mRNA | AK223474.1 | - | 4 - 4 | 1029 | 115 |
AK290193.1 | - | 5 - 5 | 1245 | 115 | |
AK291162.1 | - | 5 - 5 | 1078 | 115 | |
AK295720.1 | - | 5 - 5 | 1235 | 115 | |
AK298652.1 | - | 5 - 5 | 977 | 115 | |
AK299327.1 | - | 5 - 5 | 1034 | 115 | |
BC118586.1 | - | 5 - 5 | 1209 | 115 | |
BC121800.1 | - | 5 - 5 | 1209 | 115 | |
BG744194.1 | - | 2 - 2 | 311 | 115 | |
U90552.2 | - | 5 - 5 | 1178 | 115 | |
Y07827.1 | - | 5 - 5 | 970 | 115 |