The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003836.6 | NP_003827.3 | 5 - 5 | 1006 | 165 |
GenBank mRNA | AK289384.1 | - | 5 - 5 | 976 | 165 |
AK297918.1 | - | 4 - 4 | 902 | 165 | |
AK313258.1 | - | 5 - 5 | 976 | 165 | |
BC007741.2 | - | 5 - 5 | 955 | 165 | |
BC013197.1 | - | 5 - 5 | 922 | 165 | |
BC014015.2 | - | 5 - 5 | 951 | 165 | |
DQ309460.1 | - | 2 - 2 | 541 | 165 | |
U15979.1 | - | 5 - 5 | 975 | 165 | |
X17544.1 | - | 5 - 5 | 964 | 165 | |
Z12172.1 | - | 5 - 5 | 802 | 165 |