The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_027330.1 | 1 - 1 | 2452 | 82 | |
GenBank mRNA | AK024176.1 | - | 1 - 1 | 2364 | 82 |