Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOA
Entrez Gene ID
146183
Gene Name
otoancorin
Gene Aliases
CT108, DFNB22
Chromosome Location
Chr.16: 21663857 - 21771468 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144672.3 NP_653273.3 4 - 5 180 86
XM_011545747.2 XP_011544049.1 5 - 6 559 86
XM_017022950.1 XP_016878439.1 5 - 6 283 86
GenBank mRNA BC129992.1 - 4 - 5 177 86
BC129993.1 - 4 - 5 184 86

View Full Product Details