The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161683.1 | NP_001155155.1 | 5 - 6 | 701 | 94 |
NM_144672.3 | NP_653273.3 | 9 - 10 | 845 | 94 | |
XM_011545747.2 | XP_011544049.1 | 10 - 11 | 1224 | 94 | |
XM_017022950.1 | XP_016878439.1 | 10 - 11 | 948 | 94 | |
GenBank mRNA | AK125840.1 | - | 5 - 6 | 701 | 94 |
BC129992.1 | - | 9 - 10 | 842 | 94 | |
BC129993.1 | - | 9 - 10 | 849 | 94 |