Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
RECQL4 | NM_004260.3 | 15 | 2551 | NP_004251.3 |
XM_011517384.2 | 15 | 2353 | XP_011515686.1 | |
XM_017013991.1 | 14 | 2646 | XP_016869480.1 | |
XM_017013992.1 | 14 | 2645 | XP_016869481.1 | |
XM_017013993.1 | 15 | 2555 | XP_016869482.1 | |
XM_017013994.1 | 15 | 2551 | XP_016869483.1 | |
XM_017013995.1 | 15 | 2553 | XP_016869484.1 | |
XM_017013996.1 | 14 | 2644 | XP_016869485.1 | |
XM_017013997.1 | 14 | 2448 | XP_016869486.1 | |
XM_017013998.1 | 14 | 2643 | XP_016869487.1 | |
XM_017013999.1 | 15 | 2357 | XP_016869488.1 | |
XM_017014000.1 | 10 | 1557 | XP_016869489.1 | |
XM_017014001.1 | 13 | 2647 | XP_016869490.1 | |
AB006532.1 | 15 | 2509 | BAA74453.1 | |
BC011602.2 | 8 | 1110 | AAH11602.2 | |
BC013277.2 | 6 | 848 | AAH13277.2 | |
KJ147518.1 | 11 | 1858 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470249 | Chr8:143889098 - 144519234 on Build GRCh38 | Loss | PPP1R16A MIR6849 LOC101928953 TMEM249 SHARPIN MIR6893 VPS28 MIR939 HGH1 LRRC14 PLEC MIR661 HSF1 MIR6848 PARP10 RECQL4 SCRT1 MFSD3 WDR97 MAF1 KIFC2 SMPD5 MIR6846 GPAA1 MIR7112 TONSL-AS1 SCX EXOSC4 CYHR1 GPT MIR6847 TONSL MIR1234 MROH1 CPSF1 FOXH1 SLC39A4 OPLAH CYC1 ADCK5 GRINA FBXL6 BOP1 SLC52A2 DGAT1 LOC101928902 SPATC1 |
nsv612951 | Chr8:144479830 - 144677692 on Build GRCh38 | Loss | PPP1R16A LOC101928953 RECQL4 GPT MFSD3 C8orf82 ARHGAP39 LRRC14 LRRC24 |
nsv831489 | Chr8:144342140 - 144544026 on Build GRCh38 | Loss | PPP1R16A MIR6849 LOC101928953 TMEM249 MIR6893 VPS28 MIR939 ARHGAP39 LRRC14 LRRC24 RECQL4 MFSD3 KIFC2 TONSL-AS1 CYHR1 GPT TONSL MIR1234 C8orf82 CPSF1 FOXH1 SLC39A4 ADCK5 FBXL6 SLC52A2 LOC101928902 |
dgv1207n67 | Chr8:144498396 - 144532417 on Build GRCh38 | Gain | PPP1R16A LOC101928953 RECQL4 GPT MFSD3 C8orf82 ARHGAP39 LRRC14 LRRC24 |
dgv12439n54 | Chr8:144438306 - 144525122 on Build GRCh38 | Loss | PPP1R16A LOC101928953 RECQL4 TONSL-AS1 CYHR1 GPT MFSD3 TONSL KIFC2 LRRC14 FOXH1 LRRC24 |
dgv907n27 | Chr8:144416180 - 144525122 on Build GRCh38 | Loss | PPP1R16A LOC101928953 TONSL-AS1 MIR6893 VPS28 CYHR1 GPT TONSL LRRC14 FOXH1 LRRC24 SLC39A4 RECQL4 MFSD3 KIFC2 |
dgv12442n54 | Chr8:144492696 - 144516381 on Build GRCh38 | Loss | PPP1R16A LOC101928953 RECQL4 GPT MFSD3 |
dgv12434n54 | Chr8:144270080 - 144516801 on Build GRCh38 | Loss | PPP1R16A MIR6849 LOC101928953 TMEM249 MIR6893 VPS28 MIR939 HSF1 MIR6848 RECQL4 SCRT1 MFSD3 KIFC2 TONSL-AS1 CYHR1 GPT TONSL MIR1234 CPSF1 FOXH1 SLC39A4 ADCK5 FBXL6 BOP1 SLC52A2 DGAT1 LOC101928902 |
dgv12443n54 | Chr8:144502322 - 144518490 on Build GRCh38 | Loss | LOC101928953 RECQL4 GPT MFSD3 LRRC14 |
nsv515809 | Chr8:143904439 - 144519234 on Build GRCh38 | Gain+Loss | PPP1R16A MIR6849 LOC101928953 TMEM249 SHARPIN MIR6893 VPS28 MIR939 HGH1 LRRC14 PLEC MIR661 HSF1 MIR6848 PARP10 RECQL4 SCRT1 MFSD3 WDR97 MAF1 KIFC2 SMPD5 MIR6846 GPAA1 MIR7112 TONSL-AS1 SCX EXOSC4 CYHR1 GPT MIR6847 TONSL MIR1234 MROH1 CPSF1 FOXH1 SLC39A4 OPLAH CYC1 ADCK5 GRINA FBXL6 BOP1 SLC52A2 DGAT1 LOC101928902 SPATC1 |
nsv612940 | Chr8:144436292 - 144613673 on Build GRCh38 | Loss | PPP1R16A LOC101928953 TONSL-AS1 CYHR1 GPT TONSL C8orf82 ARHGAP39 LRRC14 FOXH1 LRRC24 RECQL4 MFSD3 KIFC2 |
nsv428208 | Chr8:144041826 - 144722291 on Build GRCh38 | Gain | PPP1R16A MIR6849 LOC101928953 TMEM249 SHARPIN MIR6893 VPS28 MIR939 ARHGAP39 HGH1 LRRC14 LRRC24 HSF1 MIR6848 RECQL4 SCRT1 MFSD3 WDR97 ZNF251 MAF1 KIFC2 SMPD5 MIR6846 GPAA1 MIR7112 TONSL-AS1 SCX EXOSC4 CYHR1 GPT MIR6847 TONSL MIR1234 MROH1 C8orf82 CPSF1 FOXH1 SLC39A4 OPLAH CYC1 ADCK5 FBXL6 BOP1 SLC52A2 DGAT1 LOC101928902 SPATC1 |
nsv612965 | Chr8:144508945 - 144516381 on Build GRCh38 | Loss | RECQL4 MFSD3 |
esv29999 | Chr8:144456732 - 144525122 on Build GRCh38 | Loss | PPP1R16A LOC101928953 RECQL4 CYHR1 GPT MFSD3 KIFC2 LRRC14 FOXH1 LRRC24 |