Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GP1BB
- Entrez Gene ID
-
2812
- Gene Name
- glycoprotein Ib platelet beta subunit
- Gene Aliases
-
BDPLT1, BS, CD42C, GPIBB, GPIbbeta
- Chromosome Location
-
Chr.22: 19723543 - 19724774 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_000407.4 |
NP_000398.1 |
2 - 2 |
913 |
178 |
GenBank mRNA |
J03259.1 |
- |
2 - 2 |
908 |
178 |
- Gene Symbol
- SEPT5-GP1BB
- Entrez Gene ID
-
100526833
- Gene Name
- SEPT5-GP1BB readthrough
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 19717220 - 19724774 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037611.1 |
|
12 - 12 |
4626 |
178 |
NR_037612.1 |
|
12 - 12 |
3130 |
178 |
GenBank mRNA |
BC024304.1 |
- |
5 - 5 |
2287 |
178 |
L20860.1 |
- |
1 - 1 |
2137 |
178 |
U59632.1 |
- |
12 - 12 |
3129 |
178 |
View Full Product Details