The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037663.1 | NP_001032752.1 | 4 - 5 | 482 | 121 |
NM_001959.3 | NP_001950.1 | 3 - 4 | 710 | 121 | |
NM_021121.3 | NP_066944.1 | 4 - 5 | 436 | 121 | |
GenBank mRNA | AK291910.1 | - | 3 - 4 | 408 | 121 |
BC000211.1 | - | 4 - 5 | 402 | 121 | |
BC004931.1 | - | 1 - 1 | 371 | 121 | |
BC029316.1 | - | 3 - 4 | 386 | 121 | |
BC067787.1 | - | 3 - 4 | 409 | 121 | |
BM747256.1 | - | 4 - 5 | 451 | 121 | |
BT007079.1 | - | 3 - 4 | 324 | 121 | |
BU627453.1 | - | 1 - 1 | 121 | ||
CR456825.1 | - | 3 - 4 | 324 | 121 | |
X60489.1 | - | 3 - 4 | 559 | 121 | |
X60656.1 | - | 4 - 5 | 419 | 121 |