Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLX1
Entrez Gene ID
1745
Gene Name
distal-less homeobox 1
Gene Aliases
-
Chromosome Location
Chr.2: 172085480 - 172089674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038493.1 NP_001033582.1 1 - 2 512 101
GenBank mRNA BC053351.1 - 1 - 2 389 101

View Full Product Details