Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA1456
Entrez Gene ID
57604
Gene Name
KIAA1456
Gene Aliases
C8orf79, TRM9L
Chromosome Location
Chr.8: 12945674 - 13029777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005273585.4 XP_005273642.1 2 - 3 349 110
XM_005273586.4 XP_005273643.1 2 - 3 352 110
XM_005273587.4 XP_005273644.1 1 - 2 253 110
XM_005273588.4 XP_005273645.1 1 - 2 379 110
XM_005273591.4 XP_005273648.1 1 - 2 379 110
XM_011544598.2 XP_011542900.1 1 - 2 419 110
XM_017013707.1 XP_016869196.1 1 - 2 379 110
XM_017013710.1 XP_016869199.1 2 - 3 325 110
XM_017013711.1 XP_016869200.1 1 - 2 247 110
XM_017013714.1 XP_016869203.1 1 - 2 379 110
GenBank mRNA AK094299.1 - 2 - 3 326 110
BC035082.1 - 1 - 2 725 110

View Full Product Details