The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020844.2 | NP_065895.2 | 2 - 3 | 486 | 76 |
XM_005273586.4 | XP_005273643.1 | 3 - 4 | 542 | 76 | |
XM_005273587.4 | XP_005273644.1 | 2 - 3 | 443 | 76 | |
XM_005273588.4 | XP_005273645.1 | 2 - 3 | 569 | 76 | |
XM_017013705.1 | XP_016869194.1 | 2 - 3 | 347 | 76 | |
XM_017013706.1 | XP_016869195.1 | 2 - 3 | 568 | 76 | |
XM_017013708.1 | XP_016869197.1 | 2 - 3 | 525 | 76 | |
XM_017013709.1 | XP_016869198.1 | 2 - 3 | 525 | 76 | |
XM_017013712.1 | XP_016869201.1 | 2 - 3 | 525 | 76 | |
XM_017013713.1 | XP_016869202.1 | 2 - 3 | 525 | 76 | |
XM_017013715.1 | XP_016869204.1 | 2 - 3 | 525 | 76 | |
GenBank mRNA | AK296010.1 | - | 3 - 4 | 339 | 76 |
BC035082.1 | - | 2 - 3 | 915 | 76 | |
DC334299.1 | - | 2 - 3 | 346 | 76 |