Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLRG2
Entrez Gene ID
346689
Gene Name
killer cell lectin like receptor G2
Gene Aliases
CLEC15B
Chromosome Location
Chr.7: 139426931 - 139483716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198508.2 NP_940910.1 4 - 5 1181 85
GenBank mRNA AK126174.1 - 4 - 5 1186 85
BC104963.1 - 4 - 5 1146 85
BC104965.1 - 4 - 5 1146 85

View Full Product Details