The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198699.1 | NP_941972.1 | 1 - 1 | 823 | 66 |
GenBank mRNA | AB076364.1 | - | 1 - 1 | 823 | 66 |