Product Details

Assay Reference Genome
Location

Chr.17:46030391 on build GRCh38
Cytoband
17q21.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
284058
Gene Symbol
KANSL1
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
Location
Chr.17:46029916-46225374 on build GRCh38
Assay Gene Location
Within Exon 20
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
KANSL1 NM_001193465.1 16 4657 NP_001180394.1
NM_001193466.1 15 4874 NP_001180395.1
NM_015443.3 15 4576 NP_056258.1
XM_006721823.1 16 4660 XP_006721886.1
XM_006721824.3 16 4844 XP_006721887.1
XM_011524628.2 16 4841 XP_011522930.1
XM_011524629.2 15 4742 XP_011522931.1
XM_011524630.2 15 4655 XP_011522932.1
XM_011524631.2 15 4653 XP_011522933.1
XM_011524632.2 15 3270 XP_011522934.1
AB033093.1 14 4497 BAA86581.1
AK094946.1 7 2375
AL117476.1 4 1744 CAB55949.1
AL137317.1 11 2818 CAB70694.1
BC035892.1 10 2681 AAH35892.1
BC098376.1 15 4576 AAH98376.1
BX538006.1 15 4877 CAD97958.1
BX648760.1 8 8634 CAH10565.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv471698 Chr17:46002113 - 46300887 on Build GRCh38 Gain+Loss KANSL1 MAPT ARL17B KANSL1-AS1 LRRC37A
nsv469705 Chr17:45969304 - 46127121 on Build GRCh38 Loss KANSL1 MAPT STH
nsv469537 Chr17:46002112 - 46163353 on Build GRCh38 Gain+Loss KANSL1 MAPT
nsv575142 Chr17:46003698 - 46172742 on Build GRCh38 Loss KANSL1 MAPT
esv2762433 Chr17:45496768 - 46710828 on Build GRCh38 Gain+Loss LRRC37A4P LRRC37A2 LOC107987254 MAPT-IT1 KANSL1 MAPT LOC107987253 ARL17B STH NSF MGC57346-CRHR1 CRHR1-IT1 LRRC37A MGC57346 NSFP1 CRHR1 LOC644172 ARL17A LOC105376839 SPPL2C MAPT-AS1 KANSL1-AS1
nsv1072740 Chr17:45970433 - 46248034 on Build GRCh38 Deletion KANSL1 MAPT STH KANSL1-AS1
dgv3178n100 Chr17:45877004 - 46034461 on Build GRCh38 Gain MAPT-IT1 KANSL1 MAPT STH MAPT-AS1
esv2751686 Chr17:45962198 - 46286690 on Build GRCh38 Gain KANSL1 MAPT ARL17B STH KANSL1-AS1 LRRC37A
esv2758692 Chr17:46006548 - 47199967 on Build GRCh38 Gain+Loss LRRC37A2 LOC107987254 KANSL1 MAPT LOC107987253 ARL17B GOSR2 NSF LRRC37A MIR5089 NSFP1 RPS2P47 ARL17A CDC27 RPRML WNT3 WNT9B KANSL1-AS1
dgv128e55 Chr17:46017105 - 46286690 on Build GRCh38 Gain KANSL1 MAPT ARL17B KANSL1-AS1 LRRC37A
dgv127e55 Chr17:45927606 - 46733750 on Build GRCh38 Gain NSFP1 LRRC37A2 LOC107987254 KANSL1 ARL17A MAPT LOC107987253 ARL17B STH NSF KANSL1-AS1 LRRC37A

View Full Product Details