Product Details

Assay Reference Genome
Location

Chr.7:100419900 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55063
Gene Symbol
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Location
Chr.7:100400826-100429239 on build GRCh38
Assay Gene Location
Overlaps Intron 4 - Exon 5
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ZCWPW1 NM_001258008.1 NP_001244937.1
NM_017984.4 NP_060454.3
XM_005250479.2 XP_005250536.1
XM_005250480.2 XP_005250537.1
XM_006716035.3 XP_006716098.1
XM_006716036.2 XP_006716099.1
XM_006716037.3 XP_006716100.1
XM_006716038.3 XP_006716101.1
XM_006716040.3 XP_006716103.1
XM_017012379.1 XP_016867868.1
XM_017012380.1 XP_016867869.1
AK093038.1 BAC04028.1
AK299989.1
AK300745.1
AK302108.1
AK316553.1
AL136735.1 CAB66669.1
BC002725.1 AAH02725.1
DB447154.1
DC413605.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion ZSCAN21 LAMTOR4 ZNF3 TAF6 SLC12A9 ZKSCAN1 FIS1 GATS MUC3A MIR25 MIR6875 ACTL6B MOSPD3 PPP1R35 MBLAC1 ZNHIT1 SERPINE1 TSC22D4 TFR2 TRIP6 PVRIG GAL3ST4 PILRB STAG3 PCOLCE-AS1 COPS6 PILRA PLOD3 MIR6840 SAP25 MUC12 MIR4653 ACHE UFSP1 NAT16 NYAP1 EPO GNB2 SPDYE3 PVRIG2P VGF PCOLCE SRRT TRIM56 AP4M1 MUC17 ZASP FBXO24 LOC102724094 ZAN LOC105375429 PMS2P1 MIR4658 AGFG2 CLDN15 MEPCE C7orf43 GIGYF1 STAG3L5P ZCWPW1 C7orf61 STAG3L5P-PVRIG2P-PILRB GPC2 AZGP1 POP7 AZGP1P1 CNPY4 MIR93 MCM7 LRCH4 EPHB4 AP1S1 MIR106B MOGAT3
nsv831071 Chr7:100320402 - 100515790 on Build GRCh38 Loss PVRIG2P PMS2P1 PILRB MEPCE PILRA STAG3L5P MIR6840 ZCWPW1 C7orf61 STAG3L5P-PVRIG2P-PILRB PPP1R35 NYAP1 SPDYE3 TSC22D4
nsv527107 Chr7:100417088 - 100467096 on Build GRCh38 Loss PPP1R35 MEPCE ZCWPW1 TSC22D4 C7orf61

View Full Product Details