Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GDAP1
Entrez Gene ID
54332
Gene Name
ganglioside induced differentiation associated protein 1
Gene Aliases
CMT4, CMT4A, CMTRIA
Chromosome Location
Chr.8: 74349251 - 74488867 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040875.2 NP_001035808.1 5 - 6 644 109
NM_018972.2 NP_061845.2 5 - 6 774 109
NR_046346.1 5 - 6 629 109
XM_011517551.2 XP_011515853.1 4 - 5 1732 109
XM_011517552.2 XP_011515854.1 4 - 5 569 109
GenBank mRNA AB551556.1 - 5 - 6 773 109
AB551557.1 - 5 - 6 773 109
AB551558.1 - 5 - 6 771 109
AK292572.1 - 5 - 6 713 109
AK295594.1 - 5 - 6 568 109
BC024939.1 - 5 - 6 609 109
BC035450.1 - 5 - 6 621 109
Y17849.1 - 5 - 6 702 109

View Full Product Details