The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005738.1 | NP_001005738.1 | 2 - 2 | 334 | 98 |
NM_001462.3 | NP_001453.1 | 2 - 2 | 432 | 98 | |
XM_006723120.3 | XP_006723183.1 | 5 - 5 | 1165 | 98 | |
XM_017026542.1 | XP_016882031.1 | 4 - 4 | 1009 | 98 | |
GenBank mRNA | AF054013.1 | - | 1 - 1 | 289 | 98 |
AK290557.1 | - | 3 - 3 | 480 | 98 | |
BC029125.2 | - | 1 - 1 | 303 | 98 | |
BC071722.1 | - | 2 - 2 | 604 | 98 | |
BM922150.1 | - | 2 - 2 | 370 | 98 | |
D10922.1 | - | 2 - 2 | 323 | 98 | |
M76672.1 | - | 1 - 1 | 289 | 98 | |
M84562.1 | - | 2 - 2 | 1061 | 98 | |
M88107.1 | - | 2 - 2 | 328 | 98 | |
U81501.1 | - | 1 - 1 | 309 | 98 | |
X63819.1 | - | 2 - 2 | 442 | 98 |