The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039999.2 | NP_001035088.2 | 5 - 5 | 1957 | 111 |
XM_017024953.1 | XP_016880442.1 | 5 - 5 | 2041 | 111 | |
GenBank mRNA | AK123558.1 | - | 3 - 3 | 2227 | 111 |
BX402942.2 | - | 1 - 1 | 671 | 111 |