The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039775.3 | NP_001034864.2 | 2 - 2 | 952 | 152 |
XM_005245918.2 | XP_005245975.1 | 2 - 2 | 938 | 152 | |
XM_011541672.1 | XP_011539974.1 | 2 - 2 | 938 | 152 | |
XM_011541673.2 | XP_011539975.1 | 2 - 2 | 1177 | 152 | |
GenBank mRNA | AK000902.1 | - | 2 - 2 | 952 | 152 |