The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006945.1 | NP_001006946.1 | 1 - 2 | 323 | 80 |
GenBank mRNA | BU664417.1 | - | 1 - 2 | 312 | 80 |
CD686306.1 | - | 1 - 2 | 361 | 80 |