Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
CYP4V2 | NM_207352.3 | NP_997235.3 | ||
XM_005262935.3 | XP_005262992.1 | |||
XM_017008037.1 | XP_016863526.1 | |||
AK022114.1 | ||||
AK122600.1 | BAC85487.1 | |||
AK126473.1 | BAC86562.1 | |||
AK311390.1 | ||||
AY422002.1 | AAR31180.1 | |||
BC041839.1 | AAH41839.1 | |||
BC060857.1 | AAH60857.1 | |||
BX648537.1 | ||||
BX648730.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3894090 | Chr4:186011626 - 186212421 on Build GRCh38 | Gain | TLR3 FLJ38576 FAM149A CYP4V2 |
nsv1028146 | Chr4:185582745 - 186417181 on Build GRCh38 | Gain | F11-AS1 F11 SORBS2 TLR3 KLKB1 FLJ38576 FAM149A CYP4V2 |
nsv4644 | Chr4:186185600 - 186219291 on Build GRCh38 | Insertion | KLKB1 FLJ38576 CYP4V2 |
nsv830172 | Chr4:186091611 - 186247270 on Build GRCh38 | Loss | KLKB1 FLJ38576 FAM149A CYP4V2 |
nsv508340 | Chr4:186138019 - 186215804 on Build GRCh38 | Deletion | KLKB1 FLJ38576 FAM149A CYP4V2 |
esv3603523 | Chr4:186176579 - 186214960 on Build GRCh38 | Gain | FLJ38576 CYP4V2 |
nsv461864 | Chr4:186085652 - 186335850 on Build GRCh38 | Gain | F11-AS1 F11 TLR3 KLKB1 FLJ38576 FAM149A CYP4V2 |
nsv596401 | Chr4:186151229 - 186445495 on Build GRCh38 | Loss | F11-AS1 F11 KLKB1 FLJ38576 FAM149A CYP4V2 |
esv3603517 | Chr4:186008703 - 186213065 on Build GRCh38 | Gain | TLR3 FLJ38576 FAM149A CYP4V2 |
dgv1052e214 | Chr4:186072002 - 186212619 on Build GRCh38 | Gain | TLR3 FLJ38576 FAM149A CYP4V2 |
nsv4642 | Chr4:186164028 - 186215260 on Build GRCh38 | Deletion | FLJ38576 FAM149A CYP4V2 |
esv3603525 | Chr4:186210472 - 186225457 on Build GRCh38 | Loss | KLKB1 CYP4V2 |