The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000107.2 | NP_000098.1 | 5 - 6 | 900 | 77 |
GenBank mRNA | AB107039.1 | - | 4 - 5 | 513 | 77 |
AK313262.1 | - | 5 - 6 | 848 | 77 | |
BC000093.2 | - | 5 - 6 | 728 | 77 | |
BT007139.1 | - | 5 - 6 | 705 | 77 | |
U18300.1 | - | 5 - 6 | 880 | 77 |