Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUCA1
Entrez Gene ID
2517
Gene Name
fucosidase, alpha-L- 1, tissue
Gene Aliases
FUCA
Chromosome Location
Chr.1: 23845077 - 23868369 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000147.4 NP_000138.2 2 - 3 606 74
XM_005245821.2 XP_005245878.1 2 - 3 743 74
XM_011541167.2 XP_011539469.1 2 - 3 191 74
XM_017000905.1 XP_016856394.1 2 - 3 288 74
GenBank mRNA AB074175.1 - 2 - 3 527 74
AK092914.1 - 3 - 4 74
AK095443.1 - 3 - 4 478 74
AK095455.1 - 3 - 4 492 74
AK225468.1 - 2 - 3 527 74
AK310143.1 - 3 - 4 589 74
AK314649.1 - 2 - 3 527 74
BC017338.2 - 2 - 3 529 74
M10355.1 - 2 - 3 301 74
M29877.1 - 2 - 3 526 74
X01390.1 - 2 - 3 301 74

View Full Product Details