The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016373.3 | NP_057457.1 | 2 - 3 | 540 | 105 |
NM_130791.3 | NP_570607.1 | 2 - 3 | 540 | 105 | |
NR_120436.1 | 2 - 3 | 654 | 105 | ||
XM_011523101.2 | XP_011521403.1 | 2 - 3 | 299 | 105 | |
XM_011523103.2 | XP_011521405.1 | 2 - 3 | 299 | 105 | |
XM_011523104.2 | XP_011521406.1 | 2 - 3 | 299 | 105 | |
XM_011523105.2 | XP_011521407.1 | 2 - 3 | 299 | 105 | |
XM_017023278.1 | XP_016878767.1 | 2 - 3 | 299 | 105 | |
GenBank mRNA | AF187015.1 | - | 2 - 3 | 258 | 105 |
AF211943.1 | - | 2 - 3 | 299 | 105 | |
AF227526.1 | - | 2 - 3 | 259 | 105 | |
AF227527.1 | - | 2 - 3 | 278 | 105 | |
AF227528.1 | - | 2 - 3 | 275 | 105 | |
AF395123.1 | - | 2 - 3 | 307 | 105 | |
AF395124.1 | - | 2 - 3 | 307 | 105 | |
AK027626.1 | - | 2 - 3 | 298 | 105 | |
AK290438.1 | - | 2 - 3 | 290 | 105 | |
AK291300.1 | - | 2 - 3 | 297 | 105 | |
AY256821.1 | - | 2 - 3 | 229 | 105 | |
BC003184.1 | - | 2 - 3 | 253 | 105 | |
BT007445.1 | - | - | 174 | 105 | |
BU157392.1 | - | 2 - 3 | 281 | 105 |