The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278288.1 | NP_001265217.1 | 1 - 2 | 222 | 87 |
NM_006916.2 | NP_008847.1 | 1 - 2 | 222 | 87 | |
GenBank mRNA | AK056028.1 | - | 1 - 2 | 159 | 87 |
DA237285.1 | - | 1 - 2 | 149 | 87 |