The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304342.1 | NP_001291271.1 | 6 - 7 | 1182 | 69 |
NM_020131.4 | NP_064516.2 | 6 - 7 | 1242 | 69 | |
XM_005245348.3 | XP_005245405.1 | 6 - 7 | 1147 | 69 | |
GenBank mRNA | AB209566.1 | - | 6 - 7 | 1115 | 69 |
AF113544.1 | - | 3 - 4 | 648 | 69 | |
AF188240.1 | - | 6 - 7 | 1396 | 69 | |
AK302895.1 | - | 6 - 7 | 1071 | 69 | |
AK314413.1 | - | 6 - 7 | 1130 | 69 | |
BC018403.2 | - | 6 - 7 | 1130 | 69 | |
BC063841.1 | - | 6 - 7 | 1142 | 69 |