The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077653.2 | NP_001071121.1 | 1 - 2 | 609 | 58 |
NM_001166220.1 | NP_001159692.1 | 1 - 2 | 609 | 58 | |
GenBank mRNA | BC120945.1 | - | 1 - 2 | 140 | 58 |
BC120946.1 | - | 1 - 2 | 140 | 58 | |
DQ986374.1 | - | 1 - 2 | 656 | 58 |