The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145230.3 | NP_660265.2 | 2 - 3 | 1253 | 100 |
NR_110612.1 | 2 - 3 | 1253 | 100 | ||
GenBank mRNA | AF452639.1 | - | 2 - 3 | 1252 | 99 |
AK172725.1 | - | 2 - 3 | 234 | 100 | |
AK290086.1 | - | 2 - 3 | 252 | 100 | |
AY037164.1 | - | 2 - 3 | 178 | 100 | |
BX640846.1 | - | 2 - 3 | 277 | 100 | |
DQ989009.1 | - | 2 - 3 | 587 | 100 |