The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077440.1 | NP_001070908.1 | 5 - 6 | 1931 | 89 |
NM_001077441.1 | NP_001070909.1 | 5 - 6 | 1418 | 89 | |
NM_001301038.1 | NP_001287967.1 | 5 - 6 | 1931 | 89 | |
NM_014739.2 | NP_055554.1 | 5 - 6 | 1937 | 89 | |
XM_005267237.3 | XP_005267294.1 | 5 - 6 | 1873 | 89 | |
XM_017011529.1 | XP_016867018.1 | 5 - 6 | 1405 | 89 | |
XM_017011530.1 | XP_016867019.1 | 5 - 6 | 2212 | 89 | |
XM_017011531.1 | XP_016867020.1 | 5 - 6 | 1348 | 89 | |
XM_017011532.1 | XP_016867021.1 | 5 - 6 | 1873 | 89 | |
GenBank mRNA | AF249273.1 | - | 3 - 4 | 1678 | 89 |
BC016682.1 | - | 5 - 6 | 1876 | 89 | |
BC047687.1 | - | 4 - 5 | 1244 | 89 | |
BC047887.1 | - | 5 - 6 | 1906 | 89 | |
BC056894.1 | - | 5 - 6 | 1918 | 89 | |
BC063846.1 | - | 5 - 6 | 1844 | 89 | |
BC078160.1 | - | 5 - 6 | 1843 | 89 | |
BC132780.1 | - | 5 - 6 | 1881 | 89 | |
BC144281.1 | - | 5 - 6 | 1875 | 89 | |
BC144299.1 | - | 5 - 6 | 1881 | 89 | |
BX648878.1 | - | 1 - 2 | 678 | 89 | |
BX649160.1 | - | 5 - 6 | 1349 | 89 | |
D79986.1 | - | 5 - 6 | 1937 | 89 |