Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
CLN8 | NM_018941.3 | 3 | 1526 | NP_061764.2 |
AF123758.1 | 3 | 1455 | AAF13116.1 | |
AF123759.1 | 3 | 1455 | AAF13117.1 | |
AF123760.1 | 3 | 1455 | AAF13118.1 | |
AF123761.1 | 3 | 1455 | AAF13119.1 | |
BC007725.2 | 3 | 1380 | AAH07725.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1024325 | Chr8:651043 - 3203837 on Build GRCh38 | Gain | LOC105377776 DLGAP2 LOC401442 MIR7160 MIR3674 KBTBD11-OT1 ARHGEF10 MIR596 LOC105377781 LOC286083 MYOM2 DLGAP2-AS1 ERICH1-AS1 LOC105377792 CLN8 LOC101928058 LOC101927815 LOC100131395 KBTBD11 ERICH1 LOC101927752 CSMD1 LOC105377777 LOC105377774 |
esv3891267 | Chr8:1755403 - 2343278 on Build GRCh38 | Gain | CLN8 LOC101928058 LOC100131395 KBTBD11 MIR7160 MIR3674 KBTBD11-OT1 LOC101927752 ARHGEF10 MIR596 LOC105377781 MYOM2 |
esv3615841 | Chr8:1705527 - 1822229 on Build GRCh38 | Loss | DLGAP2 CLN8 MIR3674 LOC101927752 MIR596 |
dgv11822n54 | Chr8:1748920 - 1829405 on Build GRCh38 | Loss | CLN8 MIR3674 LOC101927752 ARHGEF10 MIR596 |
nsv609527 | Chr8:1733567 - 1805331 on Build GRCh38 | Loss | CLN8 MIR3674 LOC101927752 |
esv3576504 | Chr8:951237 - 1906751 on Build GRCh38 | Gain | LOC105377776 ERICH1-AS1 DLGAP2 CLN8 MIR3674 LOC101927752 ARHGEF10 MIR596 LOC286083 DLGAP2-AS1 |
nsv609531 | Chr8:1768870 - 2267169 on Build GRCh38 | Gain | CLN8 LOC101928058 LOC100131395 KBTBD11 MIR7160 MIR3674 KBTBD11-OT1 ARHGEF10 MIR596 LOC105377781 MYOM2 |
nsv509244 | Chr8:1756513 - 1850704 on Build GRCh38 | Insertion | CLN8 MIR3674 LOC101927752 ARHGEF10 MIR596 |
nsv1025591 | Chr8:1755786 - 2335806 on Build GRCh38 | Gain | CLN8 LOC101928058 LOC100131395 KBTBD11 MIR7160 MIR3674 KBTBD11-OT1 LOC101927752 ARHGEF10 MIR596 LOC105377781 MYOM2 |
esv3615846 | Chr8:1735209 - 1816717 on Build GRCh38 | Loss | CLN8 MIR3674 LOC101927752 |