Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPL21
- Entrez Gene ID
-
6144
- Gene Name
- ribosomal protein L21
- Gene Aliases
-
HYPT12, L21
- Chromosome Location
-
Chr.13: 27251555 - 27256568 on Build GRCh38
- Gene Symbol
- RPL21P28
- Entrez Gene ID
-
100131205
- Gene Name
- ribosomal protein L21 pseudogene 28
- Gene Aliases
-
RPL21_8_160
- Chromosome Location
-
Chr.1: 212051486 - 212052051 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026911.1 |
|
1 - 1 |
353 |
149 |
View Full Product Details