Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM64
Entrez Gene ID
169200
Gene Name
transmembrane protein 64
Gene Aliases
-
Chromosome Location
Chr.8: 90621995 - 90645905 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008495.3 NP_001008495.2 2 - 3 943 105
GenBank mRNA AK123958.1 - 2 - 3 637 105
AL834364.1 - 2 - 3 649 105
BC113828.1 - 5 - 6 673 105

View Full Product Details