The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033930.2 | NP_001029102.1 | 4 - 5 | 410 | 109 |
NM_001321017.1 | NP_001307946.1 | 4 - 5 | 409 | 109 | |
NM_001321018.1 | NP_001307947.1 | 4 - 5 | 1036 | 109 | |
NM_001321019.1 | NP_001307948.1 | 4 - 5 | 574 | 109 | |
NM_001321020.1 | NP_001307949.1 | 4 - 5 | 608 | 109 | |
NM_001321021.1 | NP_001307950.1 | 4 - 5 | 423 | 109 | |
NM_001321022.1 | NP_001307951.1 | 3 - 4 | 323 | 109 | |
NM_003333.4 | NP_003324.1 | 4 - 5 | 388 | 109 | |
GenBank mRNA | AF348700.1 | - | 4 - 5 | 335 | 109 |
AK314717.1 | - | 4 - 5 | 320 | 109 | |
AV707734.1 | - | - | 388 | 109 | |
BC101830.1 | - | 4 - 5 | 352 | 109 | |
BC101832.1 | - | 4 - 5 | 352 | 109 | |
BC143668.1 | - | 4 - 5 | 352 | 109 | |
BE514609.1 | - | 4 - 5 | 555 | 109 | |
BG341101.1 | - | 4 - 5 | 426 | 109 | |
BQ428515.1 | - | 3 - 4 | 317 | 109 | |
BQ428688.1 | - | - | 390 | 109 | |
CB124676.1 | - | 4 - 5 | 354 | 109 | |
CD385706.1 | - | 4 - 5 | 579 | 109 | |
DA949569.1 | - | 4 - 5 | 409 | 109 | |
X56998.1 | - | 3 - 4 | 303 | 109 | |
X56999.1 | - | 3 - 4 | 312 | 109 | |
Y00361.1 | - | 2 - 3 | 178 | 109 |