The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076780.1 | NP_001070248.1 | 16 - 17 | 2784 | 78 |
NM_001278423.1 | NP_001265352.1 | 5 - 6 | 878 | 78 | |
NM_001278425.1 | NP_001265354.1 | 5 - 6 | 878 | 78 | |
NM_052892.3 | NP_443124.3 | 16 - 17 | 2784 | 78 | |
NR_126532.1 | 16 - 17 | 2784 | 78 | ||
GenBank mRNA | AB067466.1 | - | 8 - 9 | 1129 | 78 |
AK098052.1 | - | 10 - 11 | 1366 | 78 | |
AY164483.1 | - | 16 - 17 | 2784 | 78 | |
AY371495.1 | - | 5 - 6 | 840 | 78 | |
BC004562.2 | - | 5 - 6 | 845 | 78 | |
BC014157.2 | - | 5 - 6 | 845 | 78 |