Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMCO1
Entrez Gene ID
255798
Gene Name
single-pass membrane protein with coiled-coil domains 1
Gene Aliases
C3orf43
Chromosome Location
Chr.3: 196506877 - 196515366 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077657.2 NP_001071125.1 1 - 2 212 109
GenBank mRNA AK123917.1 - 1 - 2 212 109
AL833140.1 - 1 - 2 193 109
BC118556.1 - 1 - 2 174 109
BC118636.1 - 1 - 2 174 109

View Full Product Details