The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001255.2 | NP_001246.2 | 4 - 5 | 528 | 69 |
GenBank mRNA | AF099644.1 | - | 3 - 4 | 471 | 69 |
AK225504.1 | - | 4 - 5 | 507 | 69 | |
AK303844.1 | - | 5 - 6 | 483 | 69 | |
AK312780.1 | - | 4 - 5 | 507 | 69 | |
BC000624.2 | - | 3 - 4 | 535 | 69 | |
BC001088.2 | - | 4 - 5 | 528 | 69 | |
BC006272.2 | - | 3 - 4 | 470 | 69 | |
BC009425.1 | - | 4 - 5 | 510 | 69 | |
BC009426.1 | - | 4 - 5 | 510 | 69 | |
BC012803.2 | - | 4 - 5 | 510 | 69 | |
BC012827.2 | - | 4 - 5 | 528 | 69 | |
BC013303.2 | - | 4 - 5 | 510 | 69 | |
BC024257.1 | - | 4 - 5 | 510 | 69 | |
BC031294.1 | - | 4 - 5 | 510 | 69 | |
BC110321.1 | - | 3 - 4 | 491 | 69 | |
BT007388.1 | - | 3 - 4 | 427 | 69 | |
U05340.1 | - | 4 - 5 | 537 | 69 |