The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080770.1 | NP_001074239.1 | 2 - 3 | 123 | 69 |
NM_001080772.1 | NP_001074241.1 | 2 - 3 | 123 | 69 | |
NM_002255.5 | NP_002246.5 | 2 - 3 | 123 | 69 | |
GenBank mRNA | AF002979.1 | - | 2 - 3 | 83 | 69 |
AF002980.1 | - | 2 - 3 | 95 | 69 | |
AF002981.1 | - | 2 - 3 | 95 | 69 | |
AF002982.1 | - | 2 - 3 | 95 | 69 | |
AF034771.1 | - | 2 - 3 | 89 | 69 | |
AF034772.1 | - | 2 - 3 | 89 | 69 | |
AF034773.1 | - | 2 - 3 | 89 | 69 | |
AF276292.1 | - | 2 - 3 | 83 | 69 | |
AF285436.1 | - | 2 - 3 | 110 | 69 | |
AK309531.1 | - | 3 - 4 | 359 | 69 | |
AK310066.1 | - | 2 - 3 | 365 | 69 | |
AY359817.1 | - | 2 - 3 | 94 | 69 | |
AY601812.1 | - | 2 - 3 | 77 | 69 | |
AY789058.1 | - | 2 - 3 | 77 | 69 | |
AY789059.1 | - | 2 - 3 | 77 | 69 | |
AY789060.1 | - | 2 - 3 | 77 | 69 | |
AY789061.1 | - | 2 - 3 | 77 | 69 | |
AY789062.1 | - | 2 - 3 | 77 | 69 | |
BC041611.1 | - | 2 - 3 | 124 | 69 | |
BM918119.1 | - | - | 123 | 69 | |
DA590865.1 | - | 3 - 4 | 359 | 69 | |
DA934665.1 | - | 2 - 3 | 365 | 69 | |
DQ266438.1 | - | 2 - 3 | 86 | 69 | |
EU194340.1 | - | 2 - 3 | 83 | 69 | |
EU194341.1 | - | 1 - 2 | 55 | 69 | |
EU194342.1 | - | 2 - 3 | 83 | 69 | |
U71199.1 | - | 2 - 3 | 95 | 69 | |
U73394.1 | - | 2 - 3 | 95 | 69 | |
X97229.1 | - | 2 - 3 | 88 | 69 | |
X99479.1 | - | 2 - 3 | 83 | 69 | |
X99480.1 | - | 2 - 3 | 94 | 69 | |
Y13054.1 | - | 2 - 3 | 83 | 69 |