The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003671.3 | NP_003662.1 | 12 - 13 | 1797 | 101 |
XM_011519153.2 | XP_011517455.1 | 12 - 13 | 1740 | 101 | |
XM_017015247.1 | XP_016870736.1 | 12 - 13 | 1394 | 101 | |
XM_017015248.1 | XP_016870737.1 | 9 - 10 | 1153 | 101 | |
GenBank mRNA | AF023158.1 | - | 12 - 13 | 1345 | 101 |