The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030981.2 | NP_112243.1 | 2 - 3 | 183 | 68 |
XM_017018378.1 | XP_016873867.1 | 2 - 3 | 242 | 68 | |
GenBank mRNA | AF092437.1 | - | 2 - 3 | 101 | 68 |
AJ245875.1 | - | 1 - 2 | 88 | 68 | |
AK292086.1 | - | 2 - 3 | 151 | 68 | |
AK315333.1 | - | 2 - 3 | 144 | 68 | |
AL136635.1 | - | 2 - 3 | 135 | 68 | |
BC028331.1 | - | 2 - 3 | 134 | 68 | |
BC071169.1 | - | 2 - 3 | 122 | 68 | |
BG717501.1 | - | 2 - 3 | 188 | 68 |